missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4,6-dichloronicotinate, 95%

Manufacturer:  Alfa Aesar™ H6407703

 View more versions of this product

Catalog No. AAH6407703



Specifications

Specifications

Methyl 4,6-dichloronicotinate
65973-52-6
MFCD04125732
methyl 4,6-dichloronicotinate, methyl4,6-dichloronicotinate, 4,6-dichloro-nicotinic acid methyl, 4,6-dichloro-nicotinic acid methyl ester, 4,6-dichloronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 4,6-dichloro-, methyl ester, 2-p-tolylimidazo 1,2-a pyridine, pubchem12979, ksc494c6d, methyl-4,6-dichloronicotinate
COC(=O)C1=CN=C(C=C1Cl)Cl
206.022
206.02
95%
1g
C7H5Cl2NO2
13279782
JEJMDUMJSZTJTI-UHFFFAOYSA-N
methyl 4,6-dichloropyridine-3-carboxylate
2764083
Solid

Chemical Identifiers

65973-52-6
206.022
JEJMDUMJSZTJTI-UHFFFAOYSA-N
2764083
COC(=O)C1=CN=C(C=C1Cl)Cl
C7H5Cl2NO2
MFCD04125732
methyl 4,6-dichloronicotinate, methyl4,6-dichloronicotinate, 4,6-dichloro-nicotinic acid methyl, 4,6-dichloro-nicotinic acid methyl ester, 4,6-dichloronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 4,6-dichloro-, methyl ester, 2-p-tolylimidazo 1,2-a pyridine, pubchem12979, ksc494c6d, methyl-4,6-dichloronicotinate
methyl 4,6-dichloropyridine-3-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

storageNote1 : -30° to -10°C

SDS Documents