Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-(3-thienyl)benzoate, 97%

$112.55 - $424.91

Chemical Identifiers

Chemical Name or Material Methyl 4-(3-thienyl)benzoate
CAS 20608-91-7
Molecular Formula C12H10O2S
Formula Weight 218.27
MDL Number MFCD05863312
Synonym 4-(3-Thienyl)benzoic acid methyl ester; 4-(3-thienyl)benoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2738603

  SDS
alfa aesar™
H2738603
1g Each for $112.55

AAH2738606

  SDS
alfa aesar™
H2738606
5g Each for $424.91
Description & Specifications

Specifications

Formula Weight 218.27
CAS 20608-91-7
Quantity 1g
Melting Point 162° to 163°C
Synonym 4-(3-Thienyl)benzoic acid methyl ester; 4-(3-thienyl)benoic acid methyl ester
Assay 97%
Chemical Name or Material Methyl 4-(3-thienyl)benzoate
MDL Number MFCD05863312
Molecular Formula C12H10O2S