Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-(2,2,2-trimethylacetamido)nicotinate

$242.59 - $1,074.32

Chemical Identifiers

Chemical Name or Material Methyl 4-(2,2,2-trimethylacetamido)nicotinate
CAS 773134-09-1
Molecular Formula C12H16N2O3
Formula Weight 236.27
MDL Number MFCD06204163
Synonym Methyl 4-(pivaloylamino)nicotinate; 4-(2,2,2-Trimethyacetamido)pyridine-3-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5003403

  SDS
alfa aesar™
H5003403
1g Each for $242.59

AAH5003406

  SDS
alfa aesar™
H5003406
5g Each for $1,074.32
Description & Specifications

Specifications

Formula Weight 236.27
Chemical Name or Material Methyl 4-(2,2,2-trimethylacetamido)nicotinate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym Methyl 4-(pivaloylamino)nicotinate; 4-(2,2,2-Trimethyacetamido)pyridine-3-carboxylic acid methyl ester
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
MDL Number MFCD06204163
CAS 773134-09-1
Molecular Formula C12H16N2O3
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.