Please login to your online account to display your discounted pricing

Methyl 3-(trifluoromethoxy)benzoate 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 3-(trifluoromethoxy)benzoate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H7F3O3
Linear Formula CF3OC6H4CO2CH3
Formula Weight 220.15
MDL Number MFCD00041005
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC381820050

  SDS
thermo fisher scientific
AC381820050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 220.15
Quantity 5g
Color Clear colorless
Infrared Spectrum Authentic
Refractive Index 1.4430 to 1.4450 (20°C, 589nm)
Linear Formula CF3OC6H4CO2CH3
Chemical Name or Material Methyl 3-(trifluoromethoxy)benzoate
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C9H7F3O3
MDL Number MFCD00041005
CAS 148438-00-0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.