Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-(trifluoromethoxy)benzoate, 98%

$276.76 - $276.76

Chemical Identifiers

Chemical Name or Material Methyl 3-(trifluoromethoxy)benzoate
CAS 148438-00-0
Molecular Formula C9H7F3O3
Formula Weight 220.15g/mol
MDL Number MFCD00041005
Beilstein 5431823
Synonym 3-(Trifluoromethoxy)benzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0991406

 
alfa aesar™
L09914-06
5g Each for $276.76
Description & Specifications

Specifications

Boiling Point 85° to 87°C (10mmHg)
Quantity 5g
Chemical Name or Material Methyl 3-(trifluoromethoxy)benzoate
CAS 148438-00-0
Assay 98%
Molecular Formula C9H7F3O3
Formula Weight 220.15g/mol
MDL Number MFCD00041005
Beilstein 5431823
Synonym 3-(Trifluoromethoxy)benzoic acid methyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.