Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-thiopheneacetate, 98%

$55.88 - $223.19

Chemical Identifiers

Chemical Name or Material Methyl 3-thiopheneacetate
CAS 58414-52-1
Molecular Formula C7H8O2S
Formula Weight 156.2
MDL Number MFCD00052947
Beilstein 4378596
Synonym Methyl 3-thienylacetate; Thiophene-3-acetic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1249603

 
alfa aesar™
A1249603
1g Each for $55.88

AAA1249606

 
alfa aesar™
A1249606
5g Each for $223.19
Description & Specifications

Specifications

Formula Weight 156.2
Chemical Name or Material Methyl 3-thiopheneacetate
Assay 98%
Molecular Formula C7H8O2S
MDL Number MFCD00052947
CAS 58414-52-1
Beilstein 4378596
Synonym Methyl 3-thienylacetate; Thiophene-3-acetic acid methyl ester
Boiling Point 125° to 129°C (25mmHg)
Quantity 1g