Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-(thien-2-yl)acrylate, 97%

$65.73 - $65.73

Chemical Identifiers

Chemical Name or Material Methyl 3-(thien-2-yl)acrylate
CAS 20883-96-9
Molecular Formula C8H8O2S
Formula Weight 168.22
MDL Number MFCD00173772
Synonym 3-(2-Thienyl)acrylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2017703

  SDS
alfa aesar™
B2017703
1g Each for $65.73
Description & Specifications

Specifications

Formula Weight 168.22
Chemical Name or Material Methyl 3-(thien-2-yl)acrylate
Assay 97%
Molecular Formula C8H8O2S
MDL Number MFCD00173772
CAS 20883-96-9
Synonym 3-(2-Thienyl)acrylic acid methyl ester
Health Hazard 1 Warning
Melting Point 48° to 49°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.