Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-quinuclidinecarboxylate hydrochloride, 98+%

$191.36 - $589.15

Chemical Identifiers

Chemical Name or Material Methyl 3-quinuclidinecarboxylate hydrochloride
CAS 54954-73-3
Molecular Formula C9H15NO2·HCl
Formula Weight 205.69
MDL Number MFCD01073582
Beilstein 4272456
Synonym 3-Quinuclidinecarboxylic acid methyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1913203

  SDS
alfa aesar™
L1913203
1g Each for $589.15

AAL19132MD

  SDS
alfa aesar™
L19132MD
250mg Each for $191.36
Description & Specifications

Specifications

Formula Weight 205.69
Chemical Name or Material Methyl 3-quinuclidinecarboxylate hydrochloride
Assay ≥98%
MDL Number MFCD01073582
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C9H15NO2·HCl
CAS 54954-73-3
Beilstein 4272456
Synonym 3-Quinuclidinecarboxylic acid methyl ester hydrochloride
Health Hazard 1 Warning
Melting Point ≈160°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.