missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-phenylpropionate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0531030

 View more versions of this product

Catalog No. AAL0531030


Description

Description

Methyl 3-phenylpropionate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Phenylpropionic Acid Methyl Ester is an ester derivative of 3-phenylpropionic acid that is sometimes present in fragrance ingredients. It is also used as an application, aromatic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-phenylpropionate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Phenylpropionic Acid Methyl Ester is an ester derivative of 3-phenylpropionic acid that is sometimes present in fragrance ingredients. It is also used as an application, aromatic intermediate.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Specifications

Specifications

Methyl 3-phenylpropionate
1.045
100°C (212°F)
1.502
250 g
methyl 3-phenylpropionate, methyl hydrocinnamate, benzenepropanoic acid, methyl ester, 3-phenylpropionic acid methyl ester, methyl benzenepropanoate, hydrocinnamic acid, methyl ester, methyl dihydrocinnamate, methyl beta-phenylpropionate, methyl b-phenylpropionate, unii-111lc1gi10
RPUSRLKKXPQSGP-UHFFFAOYSA-N
methyl 3-phenylpropanoate
7643
98%
103-25-3
236°C to 239°C
C10H12O2
MFCD00017209
1909591
Soluble in chloroform. Insoluble in water.
COC(=O)CCC1=CC=CC=C1
164.204
164.2

Chemical Identifiers

103-25-3
164.204
RPUSRLKKXPQSGP-UHFFFAOYSA-N
7643
COC(=O)CCC1=CC=CC=C1
C10H12O2
MFCD00017209
methyl 3-phenylpropionate, methyl hydrocinnamate, benzenepropanoic acid, methyl ester, 3-phenylpropionic acid methyl ester, methyl benzenepropanoate, hydrocinnamic acid, methyl ester, methyl dihydrocinnamate, methyl beta-phenylpropionate, methyl b-phenylpropionate, unii-111lc1gi10
methyl 3-phenylpropanoate
Safety and Handling

Safety and Handling

EINECSNumber : 203-092-2

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only