missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-oxotetrahydrothiophene-2-carboxylate, 98%

Manufacturer:  Alfa Aesar™ 4363403

 View more versions of this product

Catalog No. AA4363403



Specifications

Specifications

Methyl 3-oxotetrahydrothiophene-2-carboxylate
2689-69-2
MFCD00156156
TUSSXVYKOOVOID-UHFFFAOYSA-N
methyl 3-oxothiolane-2-carboxylate
102870
Liquid
1g
C6H8O3S
2-carbomethoxy-2,3,4,5-tetrahydrothiophen-3-one
COC(=O)C1C(=O)CCS1
160.187
160.19
98%

Chemical Identifiers

2689-69-2
160.187
TUSSXVYKOOVOID-UHFFFAOYSA-N
102870
COC(=O)C1C(=O)CCS1
C6H8O3S
MFCD00156156
2-carbomethoxy-2,3,4,5-tetrahydrothiophen-3-one
methyl 3-oxothiolane-2-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents