Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-oxotetrahydrothiophene-2-carboxylate, 98%

$85.58 - $970.35

Chemical Identifiers

Chemical Name or Material Methyl 3-oxotetrahydrothiophene-2-carboxylate
CAS 2689-69-2
Molecular Formula C6H8O3S
Formula Weight 160.19
MDL Number MFCD00156156
Synonym 3-Oxotetrahydrothiophene-2-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4363406

  SDS
alfa aesar™
4363406
5g Each for $970.35

AA43634MD

  SDS
alfa aesar™
43634MD
250mg Each for $85.58
Description & Specifications

Specifications

Formula Weight 160.19
Chemical Name or Material Methyl 3-oxotetrahydrothiophene-2-carboxylate
Physical Form Liquid
Assay 98%
Molecular Formula C6H8O3S
MDL Number MFCD00156156
CAS 2689-69-2
Synonym 3-Oxotetrahydrothiophene-2-carboxylic acid methyl ester
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.