missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-oxo-pentanoate, 99+%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 243160250

 View more versions of this product

Catalog No. AC243160250



Description

Description

Specifications

Specifications

Methyl 3-oxo-pentanoate
71°C
Glass bottle
1.03
Colorless to Yellow
25mL
30414-53-0
C6H10O3
MFCD00011705
3-keto-n-valeric acid methyl ester, 3-oxo-pentanoic acid methyl ester, 3-oxovaleric acid methyl ester, methyl 3-oxo-n-valerate, methyl 3-oxo-pentanoate, methyl 3-oxovalerate, methyl propionylacetate, methyl-3-oxo pentanoate, pentanoic acid, 3-oxo-, methyl ester, valeric acid, 3-oxo-, methyl ester
XJMIXEAZMCTAGH-UHFFFAOYSA-N
methyl 3-oxopentanoate
121699
Liquid
1.0300g/mL
Authentic
1.4220 to 1.424
95°C to 97°C (48.0mmHg)
-35°C
99+%
99+%
CH3CH2COCH2COOCH3
10,415; 01,1022; 04,437; 05,589; 07,324; 11,472; 12,126; 16,303
Solubility in water: slightly soluble. Other solubilities: miscible with most common organic solvents
CCC(=O)CC(=O)OC
130.15
130.15
99+%

Chemical Identifiers

30414-53-0
130.15
XJMIXEAZMCTAGH-UHFFFAOYSA-N
121699
CCC(=O)CC(=O)OC
C6H10O3
MFCD00011705
3-keto-n-valeric acid methyl ester, 3-oxo-pentanoic acid methyl ester, 3-oxovaleric acid methyl ester, methyl 3-oxo-n-valerate, methyl 3-oxo-pentanoate, methyl 3-oxovalerate, methyl propionylacetate, methyl-3-oxo pentanoate, pentanoic acid, 3-oxo-, methyl ester, valeric acid, 3-oxo-, methyl ester
methyl 3-oxopentanoate
Safety and Handling

Safety and Handling

EINECSNumber : 250-184-3

SDS Documents