missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 3-oxo-pentanoate, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 243161000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl 3-oxo-pentanoate | |
30414-53-0 | |
95°C to 97°C (48.0mmHg) | |
Authentic | |
Glass bottle | |
1.4220 to 1.424 | |
MFCD00011705 | |
10,415; 01,1022; 04,437; 05,589; 07,324; 11,472; 12,126; 16,303 | |
methyl 3-oxovalerate, methyl propionylacetate, methyl 3-oxo-pentanoate, pentanoic acid, 3-oxo-, methyl ester, methyl-3-oxo pentanoate, methyl 3-oxo-n-valerate, valeric acid, 3-oxo-, methyl ester, 3-oxovaleric acid methyl ester, 3-oxo-pentanoic acid methyl ester, 3-keto-n-valeric acid methyl ester | |
XJMIXEAZMCTAGH-UHFFFAOYSA-N | |
methyl 3-oxopentanoate | |
121699 | |
99+% |
99+% | |
1.0300g/mL | |
71°C | |
99+% | |
C6H10O3 | |
CH3CH2COCH2COOCH3 | |
100mL | |
1.03 | |
Solubility in water: slightly soluble. Other solubilities: miscible with most common organic solvents | |
CCC(=O)CC(=O)OC | |
130.14 | |
130.15 |
Safety and Handling
EINECSNumber : 250-184-3
RUO â Research Use Only