missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-nitrobenzoate, 98+%

$27.08 - $460.24

Chemical Identifiers

CAS 618-95-1
Molecular Formula C8H7NO4
Formula Weight 181.15
MDL Number MFCD00007250
Beilstein 392449
Synonym 3-Nitrobenzoic acid methyl ester
Chemical Name or Material Methyl 3-nitrobenzoate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA151550B SDS Alfa Aesar™
A151550B
1000g Each for $460.24
AAA1515518 SDS Alfa Aesar™
A1515518
50g Each for $27.08
AAA1515530 SDS Alfa Aesar™
A1515530
250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Methyl 3-nitrobenzoate was used in the preparation of iodoarene.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Description & Specifications

Chemical Identifiers

618-95-1
181.15
392449
Methyl 3-nitrobenzoate
C8H7NO4
MFCD00007250
3-Nitrobenzoic acid methyl ester

Specifications

618-95-1
181.15
392449
77° to 80°C
1000g
≥98%
C8H7NO4
MFCD00007250
3-Nitrobenzoic acid methyl ester
279°C
Methyl 3-nitrobenzoate

Safety and Handling

TSCA : TSCA