missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-nitrobenzoate, 98+%

Manufacturer:  Alfa Aesar™ A151550B

 View more versions of this product

Catalog No. AAA151550B



Description

Description

Applications
Methyl 3-nitrobenzoate was used in the preparation of iodoarene.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

Methyl 3-nitrobenzoate
77°C to 80°C
618-95-1
MFCD00007250
3-nitrobenzoic acid methyl ester, methyl m-nitrobenzoate, methyl3-nitrobenzoate, benzoic acid, 3-nitro-, methyl ester, m-nitrobenzoic acid, methyl ester, benzoic acid, m-nitro-, methyl ester, 3-nitro-benzoic acid methyl ester, 3-nitro-benzoicacimethylester, acmc-209mxc, methyl 3-nitro-benzoate
COC(=O)C1=CC(=CC=C1)[N+](=O)[O-]
181.147
181.15
279°C
1000g
C8H7NO4
392449
AXLYJLKKPUICKV-UHFFFAOYSA-N
methyl 3-nitrobenzoate
69260
≥98%

Chemical Identifiers

618-95-1
181.147
AXLYJLKKPUICKV-UHFFFAOYSA-N
69260
COC(=O)C1=CC(=CC=C1)[N+](=O)[O-]
C8H7NO4
MFCD00007250
3-nitrobenzoic acid methyl ester, methyl m-nitrobenzoate, methyl3-nitrobenzoate, benzoic acid, 3-nitro-, methyl ester, m-nitrobenzoic acid, methyl ester, benzoic acid, m-nitro-, methyl ester, 3-nitro-benzoic acid methyl ester, 3-nitro-benzoicacimethylester, acmc-209mxc, methyl 3-nitro-benzoate
methyl 3-nitrobenzoate
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents