Please login to your online account to display your discounted pricing

Methyl 3-(methylthio)propionate, 99+%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  147001000

 View more versions of this product

Catalog No. AC147001000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 134.19
Quantity 100g
Color Clear colorless
EINECS Number 236-883-6
TSCA TSCA
Density 1.0700g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4640 to 1.4660 (20°C, 589nm)
Chemical Name or Material Methyl 3-(methylthio)propionate
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C5H10O2S
Linear Formula CH3SCH2CH2CO2CH3
MDL Number MFCD00008565
CAS 13532-18-8
Health Hazard 1 STENCH

Specifications

Chemical Name or Material Methyl 3-(methylthio)propionate
Name Note 99+%
CAS 13532-18-8
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C5H10O2S
Linear Formula CH3SCH2CH2CO2CH3
Formula Weight 134.19
MDL Number MFCD00008565
Accessories

STENCH

SDS & Safety Documents