Please login to your online account to display your discounted pricing

Methyl 3-(methylthio)propionate, 99+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  147000250

 View more versions of this product

Catalog No. AC147000250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 134.19
Quantity 25g
Color Clear colorless
EINECS Number 236-883-6
TSCA TSCA
Density 1.0700g/mL
Chemical Name or Material Methyl 3-(methylthio)propionate
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99+%
Refractive Index 1.4640 to 1.4660 (20°C, 589nm)
Assay Percent Range 99+%
Linear Formula CH3SCH2CH2CO2CH3
MDL Number MFCD00008565
Health Hazard 1 STENCH
Molecular Formula C5H10O2S
CAS 13532-18-8

Specifications

Chemical Name or Material Methyl 3-(methylthio)propionate
Name Note 99+%
CAS 13532-18-8
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C5H10O2S
Linear Formula CH3SCH2CH2CO2CH3
Formula Weight 134.19
MDL Number MFCD00008565
Accessories

STENCH

SDS & Safety Documents