Please login to your online account to display your discounted pricing

Methyl 3-methylphenylacetate, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC374080250

 View more versions of this product

Catalog No. AC374080250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 164.2
Physical Form Liquid
Quantity 25g
Color Clear colorless to slightly yellow
Refractive Index 1.5045 to 1.5065 (20°C, 589nm)
MDL Number MFCD00968468
Assay Percent Range 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3C6H4CH2COOCH3
Chemical Name or Material Methyl 3-methylphenylacetate
CAS 53088-69-0
Molecular Formula C10H12O2

Specifications

Chemical Name or Material Methyl 3-methylphenylacetate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H12O2
Linear Formula CH3C6H4CH2COOCH3
Formula Weight 164.2
MDL Number MFCD00968468