Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-methylbenzo[b]thiophene-2-carboxylate, 97%

$761.95 - $761.95

Chemical Identifiers

Chemical Name or Material Methyl 3-methylbenzo[b]thiophene-2-carboxylate
CAS 3133-81-1
Molecular Formula C11H10O2S
Formula Weight 206.26
MDL Number MFCD02683110
Synonym 3-Methylbenzo[b]thiophene-2-carboxylic acid methyl ester; Methyl 3-methylthianaphthene-2-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2406914

 
alfa aesar™
B2406914
25g Each for $761.95
Description & Specifications

Specifications

Formula Weight 206.26
Chemical Name or Material Methyl 3-methylbenzo[b]thiophene-2-carboxylate
Assay 97%
Molecular Formula C11H10O2S
MDL Number MFCD02683110
CAS 3133-81-1
Synonym 3-Methylbenzo[b]thiophene-2-carboxylic acid methyl ester; Methyl 3-methylthianaphthene-2-carboxylate
Melting Point 102° to 105°C
Quantity 25g