missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-Methoxypropionate, TCI America™

$56.53 - $209.31

Chemical Identifiers

CAS 3852-09-3
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00059085
InChI Key BDJSOPWXYLFTNW-UHFFFAOYSA-N
Synonym 3-Methoxypropionic Acid Methyl Ester
PubChem CID 19710
IUPAC Name methyl 3-methoxypropanoate
SMILES COCCC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M096225ML View Documents TCI America
M096225ML
25mL Each for $56.53
 
M0962500ML View Documents TCI America
M0962500ML
500mL Each for $209.31
 
Specifications

Chemical Identifiers

3852-09-3
118.13
BDJSOPWXYLFTNW-UHFFFAOYSA-N
19710
COCCC(=O)OC
C5H10O3
MFCD00059085
3-Methoxypropionic Acid Methyl Ester
methyl 3-methoxypropanoate

Specifications

144°C
25mL
C5H10O3
3272
BDJSOPWXYLFTNW-UHFFFAOYSA-N
methyl 3-methoxypropanoate
19710
Clear Liquid at 20°C
Methyl 3-Methoxypropionate
Colorless
3852-09-3
MFCD00059085
3-Methoxypropionic Acid Methyl Ester
COCCC(=O)OC
118.13
118.13
≥99.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1350

RTECSNumber : UF5274000

TSCA : Yes