missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 3-Methoxyisobutyrate 98.0+%, TCI America™
$59.73 - $169.22
Chemical Identifiers
CAS | 3852-11-7 |
---|---|
Molecular Formula | C6H12O3 |
Molecular Weight (g/mol) | 132.16 |
MDL Number | MFCD00671553 |
InChI Key | LYLUAHKXJUQFDG-UHFFFAOYNA-N |
Synonym | 3-Methoxyisobutyric Acid Methyl Ester, Methyl 3-Methoxy-2-methylpropionate, 3-Methoxy-2-methylpropionic Acid Methyl Ester |
PubChem CID | 107177 |
IUPAC Name | methyl 3-methoxy-2-methylpropanoate |
SMILES | CC(COC)C(=O)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
M119125ML
|
TCI America
M119125ML |
25 mL |
Each for $59.73
|
|
|||||
M1191500ML
|
TCI America
M1191500ML |
500 mL |
Each for $169.22
|
|
|||||
Chemical Identifiers
3852-11-7 | |
132.16 | |
LYLUAHKXJUQFDG-UHFFFAOYNA-N | |
107177 | |
CC(COC)C(=O)OC |
C6H12O3 | |
MFCD00671553 | |
3-Methoxyisobutyric Acid Methyl Ester, Methyl 3-Methoxy-2-methylpropionate, 3-Methoxy-2-methylpropionic Acid Methyl Ester | |
methyl 3-methoxy-2-methylpropanoate |
Specifications
3852-11-7 | |
148°C | |
MFCD00671553 | |
1993 | |
LYLUAHKXJUQFDG-UHFFFAOYNA-N | |
methyl 3-methoxy-2-methylpropanoate | |
107177 | |
≥98.0% (GC) | |
Methyl 3-Methoxyisobutyrate |
Colorless | |
C6H12O3 | |
25 mL | |
3-Methoxyisobutyric Acid Methyl Ester, Methyl 3-Methoxy-2-methylpropionate, 3-Methoxy-2-methylpropionic Acid Methyl Ester | |
CC(COC)C(=O)OC | |
132.16 | |
132.16 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-1405
TSCA : No