missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-Methoxyacrylate, TCI America™

$41.25 - $325.42

Chemical Identifiers

CAS 34846-90-7
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00128206
InChI Key AUTCCPQKLPMHDN-ONEGZZNKSA-N
Synonym methyl 3-methoxyacrylate, methyl trans-3-methoxyacrylate, methyl 2e-3-methoxyprop-2-enoate, e-methyl 3-methoxyacrylate, 3-methoxyacrylic acid methyl ester, methyl-3-methoxyacrylate, 2-propenoic acid, 3-methoxy-, methyl ester, 2e, methyl trans-3-methoxy acrylate, e-methyl-3-methoxyacrylate, methyl-trans-3-methoxyacrylate
PubChem CID 5323651
IUPAC Name methyl (E)-3-methoxyprop-2-enoate
SMILES COC=CC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M099525ML View Documents TCI America
M099525ML
25mL Each for $41.25
 
M0995500ML View Documents TCI America
M0995500ML
500mL Each for $325.42
 
Specifications

Chemical Identifiers

34846-90-7
116.116
AUTCCPQKLPMHDN-ONEGZZNKSA-N
5323651
COC=CC(=O)OC
C5H8O3
MFCD00128206
methyl 3-methoxyacrylate, methyl trans-3-methoxyacrylate, methyl 2e-3-methoxyprop-2-enoate, e-methyl 3-methoxyacrylate, 3-methoxyacrylic acid methyl ester, methyl-3-methoxyacrylate, 2-propenoic acid, 3-methoxy-, methyl ester, 2e, methyl trans-3-methoxy acrylate, e-methyl-3-methoxyacrylate, methyl-trans-3-methoxyacrylate
methyl (E)-3-methoxyprop-2-enoate

Specifications

56°C
25mL
C5H8O3
methyl 3-methoxyacrylate, methyl trans-3-methoxyacrylate, methyl 2e-3-methoxyprop-2-enoate, e-methyl 3-methoxyacrylate, 3-methoxyacrylic acid methyl ester, methyl-3-methoxyacrylate, 2-propenoic acid, 3-methoxy-, methyl ester, 2e, methyl trans-3-methoxy acrylate, e-methyl-3-methoxyacrylate, methyl-trans-3-methoxyacrylate
COC=CC(=O)OC
116.116
116.12
≥95.0% (GC)
Yellow
34846-90-7
MFCD00128206
AUTCCPQKLPMHDN-ONEGZZNKSA-N
methyl (E)-3-methoxyprop-2-enoate
5323651
Clear Liquid at 20°C
Methyl 3-Methoxyacrylate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3565

TSCA : No