missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 3-methoxy-2-nitrobenzoate, 98%, Thermo Scientific Chemicals
$274.32 - $274.32
Chemical Identifiers
CAS | 5307-17-5 |
---|---|
Molecular Formula | C9H9NO5 |
Molecular Weight (g/mol) | 211.17 |
MDL Number | MFCD00051968 |
InChI Key | FDQQRLPHAAICCR-UHFFFAOYSA-N |
Synonym | benzoic acid, 3-methoxy-2-nitro-, methyl ester, 3-methoxy-2-nitrobenzoic acid methyl ester, m-anisic acid, 2-nitro-, methyl ester, maybridge1_002124, cbmicro_008502, methyl 2-nitro-m-anisate, methyl 3-methoxy-2-nitro-benzoate, 2-nitro-m-anisic acid methyl ester, 3-methoxy-2-nitro-benzoic acid methyl ester |
PubChem CID | 79195 |
IUPAC Name | methyl 3-methoxy-2-nitrobenzoate |
SMILES | COC(=O)C1=CC=CC(OC)=C1[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0888606
|
Thermo Scientific Chemicals
L0888606 |
5 g |
N/A
|
|
|||||
Description
Methyl 3-methoxy-2-nitrobenzoate is used as Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 3-methoxy-2-nitrobenzoate is used as Pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store away from oxidizing agents.
Chemical Identifiers
5307-17-5 | |
211.17 | |
FDQQRLPHAAICCR-UHFFFAOYSA-N | |
79195 | |
COC(=O)C1=CC=CC(OC)=C1[N+]([O-])=O |
C9H9NO5 | |
MFCD00051968 | |
benzoic acid, 3-methoxy-2-nitro-, methyl ester, 3-methoxy-2-nitrobenzoic acid methyl ester, m-anisic acid, 2-nitro-, methyl ester, maybridge1_002124, cbmicro_008502, methyl 2-nitro-m-anisate, methyl 3-methoxy-2-nitro-benzoate, 2-nitro-m-anisic acid methyl ester, 3-methoxy-2-nitro-benzoic acid methyl ester | |
methyl 3-methoxy-2-nitrobenzoate |
Specifications
5307-17-5 | |
C9H9NO5 | |
5 g | |
benzoic acid, 3-methoxy-2-nitro-, methyl ester, 3-methoxy-2-nitrobenzoic acid methyl ester, m-anisic acid, 2-nitro-, methyl ester, maybridge1_002124, cbmicro_008502, methyl 2-nitro-m-anisate, methyl 3-methoxy-2-nitro-benzoate, 2-nitro-m-anisic acid methyl ester, 3-methoxy-2-nitro-benzoic acid methyl ester | |
FDQQRLPHAAICCR-UHFFFAOYSA-N | |
methyl 3-methoxy-2-nitrobenzoate | |
79195 | |
98% |
142°C to 144°C | |
MFCD00051968 | |
2697098 | |
Insoluble in water. | |
COC(=O)C1=CC=CC(OC)=C1[N+]([O-])=O | |
211.17 | |
211.18 | |
Methyl 3-methoxy-2-nitrobenzoate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only