missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-mercaptopropionate 98%, ACROS Organics™

Manufacturer:  Acros Organics 148875000

 View more versions of this product

Catalog No. AC148875000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methyl 3-mercaptopropionate
60°C
Glass bottle
1.08
Colorless
98%
98%
HSCH2CH2CO2CH3
03, II, 214
LDTLDBDUBGAEDT-UHFFFAOYSA-N
methyl 3-sulfanylpropanoate
18050
Liquid
1.0800g/mL
Authentic
1.4620 to 1.4640
166.0°C to 169.0°C
500g
2935-90-2
C4H8O2S
MFCD00004895
methyl 3-mercaptopropionate, propanoic acid, 3-mercapto-, methyl ester, 3-mercaptopropionic acid methyl ester, methyl mercaptopropionate, methyl3-mercaptopropionate, methyl 3-mercaptopropanoate, propionic acid, 3-mercapto-, methyl ester, unii-v920f0bcrz, methyl-3-mercaptopropionate, v920f0bcrz
COC(=O)CCS
120.166
120.17
98%

Chemical Identifiers

2935-90-2
120.166
LDTLDBDUBGAEDT-UHFFFAOYSA-N
18050
COC(=O)CCS
C4H8O2S
MFCD00004895
methyl 3-mercaptopropionate, propanoic acid, 3-mercapto-, methyl ester, 3-mercaptopropionic acid methyl ester, methyl mercaptopropionate, methyl3-mercaptopropionate, methyl 3-mercaptopropanoate, propionic acid, 3-mercapto-, methyl ester, unii-v920f0bcrz, methyl-3-mercaptopropionate, v920f0bcrz
methyl 3-sulfanylpropanoate
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Toxic if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

GHS Signal Word: Danger

EINECSNumber : 220-912-4

TSCA : TSCA

SDS Documents