Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-isothiocyanatopropionate, 99%

Chemical Identifiers

Chemical Name or Material Methyl 3-isothiocyanatopropionate
CAS 18967-35-6
Molecular Formula C5H7NO2S
Linear Formula SCN(CH2)2CO2CH3
Formula Weight 145.18
MDL Number MFCD00041129
UN Number UN2922
Beilstein 1704701
Synonym 3-Isothiocyanatopropionic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0999603

  SDS
alfa aesar™
L0999603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0999606

  SDS
alfa aesar™
L0999606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 145.18
CAS 18967-35-6
Molecular Formula C5H7NO2S
Chemical Name or Material Methyl 3-isothiocyanatopropionate
MDL Number MFCD00041129
Synonym 3-Isothiocyanatopropionic acid methyl ester
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
UN Number UN2922
Odor Irritating
Health Hazard 1 Danger
Assay 99%
Linear Formula SCN(CH2)2CO2CH3
Recommended Storage 2° to 8°C
Boiling Point 91° to 92°C (10mmHg)
Quantity 1g
Beilstein 1704701
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

2° to 8°C