missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-isothiocyanatopropionate, 99%

Manufacturer:  Alfa Aesar™ L0999606

 View more versions of this product

Catalog No. AAL0999606



Specifications

Specifications

Methyl 3-isothiocyanatopropionate
91°C to 92°C (10mmHg)
18967-35-6
SCN(CH2)2CO2CH3
UN2922
methyl 3-isothiocyanatopropionate, acmc-20akh7, methyl-3-isothiocyanatopropionate, 2-methoxycarbonylethylisothiocyanate, methyl3-isothiocyanatopropionate, 3-isothiocyanatopropanoic acid methyl ester, 3-isothiocyanatopropionic acid methyl ester, 3-isothiocyanato-propionic acid methyl ester, propanoic acid,3-isothiocyanato-, methyl ester
COC(=O)CCN=C=S
145.176
145.18
Irritating
5g
C5H7NO2S
MFCD00041129
1704701
VLIJIUDWAKBKSO-UHFFFAOYSA-N
methyl 3-isothiocyanatopropanoate
140454
99%

Chemical Identifiers

18967-35-6
145.176
VLIJIUDWAKBKSO-UHFFFAOYSA-N
140454
COC(=O)CCN=C=S
C5H7NO2S
MFCD00041129
methyl 3-isothiocyanatopropionate, acmc-20akh7, methyl-3-isothiocyanatopropionate, 2-methoxycarbonylethylisothiocyanate, methyl3-isothiocyanatopropionate, 3-isothiocyanatopropanoic acid methyl ester, 3-isothiocyanatopropionic acid methyl ester, 3-isothiocyanato-propionic acid methyl ester, propanoic acid,3-isothiocyanato-, methyl ester
methyl 3-isothiocyanatopropanoate
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents