Please login to your online account to display your discounted pricing

Methyl 3-isocyanatobenzoate, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  AC31920DE

 View more versions of this product

Catalog No. AC31920DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 177.16
Quantity 5g
Chemical Name or Material Methyl 3-isocyanatobenzoate
Assay Percent Range 97%
Molecular Formula C9H7NO3
CAS 41221-47-0
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material Methyl 3-isocyanatobenzoate
Assay Percent Range 97%
Molecular Formula C9H7NO3
Formula Weight 177.16