missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-isocyanatobenzoate, 97%, Maybridge

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C9H7NO3
Formula Weight 177.16
Chemical Name or Material Methyl 3-isocyanatobenzoate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC31920DE Fisher Scientific Company
AC31920DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC31920EA SDS Fisher Scientific Company
AC31920EA
Amber Glass Bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC31920DA SDS Fisher Scientific Company
AC31920DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
C9H7NO3
177.16
Methyl 3-isocyanatobenzoate

Specifications

177.16
41221-47-0
C9H7NO3
Methyl 3-isocyanatobenzoate
5g
Amber Glass Bottle
97%