Please login to your online account to display your discounted pricing

Methyl 3-iodo-1H-indole-6-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 3-iodo-1H-indole-6-carboxylate
CAS 850374-98-0
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H8INO2
Formula Weight 301.08
MDL Number MFCD06797504
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07519CB

  SDS
fisher scientific company
MO07519CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 301.08
Quantity 250mg
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
CAS 850374-98-0
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Methyl 3-iodo-1H-indole-6-carboxylate
Packaging Amber Glass Bottle
Molecular Formula C10H8INO2
MDL Number MFCD06797504