missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-Hydroxyphenylacetate, TCI America™

$96.25 - $337.64

Chemical Identifiers

CAS 42058-59-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00017660
InChI Key AMDDOQIUPAINLH-UHFFFAOYSA-N
Synonym 3-Hydroxyphenylacetic Acid Methyl Ester
PubChem CID 39129
IUPAC Name methyl 2-(3-hydroxyphenyl)acetate
SMILES COC(=O)CC1=CC(=CC=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M29251G View Documents TCI America
M29251G
1g Each for $96.25
 
M29255G View Documents TCI America
M29255G
5g Each for $337.64
 
Specifications

Chemical Identifiers

42058-59-3
166.176
AMDDOQIUPAINLH-UHFFFAOYSA-N
39129
COC(=O)CC1=CC(=CC=C1)O
C9H10O3
MFCD00017660
3-Hydroxyphenylacetic Acid Methyl Ester
methyl 2-(3-hydroxyphenyl)acetate

Specifications

146°C
1g
C9H10O3
3-Hydroxyphenylacetic Acid Methyl Ester
COC(=O)CC1=CC(=CC=C1)O
166.176
166.18
≥98.0% (GC)
Yellow
42058-59-3
MFCD00017660
AMDDOQIUPAINLH-UHFFFAOYSA-N
methyl 2-(3-hydroxyphenyl)acetate
39129
Clear Liquid at 20°C
Methyl 3-Hydroxyphenylacetate
Safety and Handling

Safety and Handling

TSCA : No