Please login to your online account to display your discounted pricing

Methyl 3-hydroxybenzoate, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  352001000

 View more versions of this product

Catalog No. AC352001000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 152.15
Physical Form Crystals
Assay ≥97.5%
Quantity 100g
Color Beige
EINECS Number 243-071-5
Melting Point 70° to 74°C
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Linear Formula HOC6H4CO2CH3
MDL Number MFCD00002295
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material Methyl 3-hydroxybenzoate
Molecular Formula C8H8O3
CAS 19438-10-9

Specifications

Chemical Name or Material Methyl 3-hydroxybenzoate
Name Note 98%
CAS 19438-10-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H8O3
Linear Formula HOC6H4CO2CH3
Formula Weight 152.15
MDL Number MFCD00002295
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents