missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-hydroxy-4-nitrobenzoate, 98%

Manufacturer:  Alfa Aesar™ A1532922

 View more versions of this product

Catalog No. AAA1532922



Specifications

Specifications

Methyl 3-hydroxy-4-nitrobenzoate
100g
C8H7NO5
2583832
UEGCRFNWTGYVKX-UHFFFAOYSA-N
methyl 3-hydroxy-4-nitrobenzoate
294866
98%
89°C to 92°C
713-52-0
MFCD00016997
benzoic acid, 3-hydroxy-4-nitro-, methyl ester, 3-hydroxy-4-nitrobenzoic acid methyl ester, 3-hydroxy-4-nitro-benzoic acid methyl ester, methyl 3-hydroxy-4-nitro-benzoate, methyl3-hydroxy-4-nitrobenzoate, pubchem20121, acmc-209oia, ksc379e5n, 5-methoxycarbonyl-2-nitrophenol, methyl3-hydroxy-4-nitronemzoate
COC(=O)C1=CC(=C(C=C1)[N+](=O)[O-])O
197.146
197.15

Chemical Identifiers

713-52-0
197.146
UEGCRFNWTGYVKX-UHFFFAOYSA-N
294866
COC(=O)C1=CC(=C(C=C1)[N+](=O)[O-])O
C8H7NO5
MFCD00016997
benzoic acid, 3-hydroxy-4-nitro-, methyl ester, 3-hydroxy-4-nitrobenzoic acid methyl ester, 3-hydroxy-4-nitro-benzoic acid methyl ester, methyl 3-hydroxy-4-nitro-benzoate, methyl3-hydroxy-4-nitrobenzoate, pubchem20121, acmc-209oia, ksc379e5n, 5-methoxycarbonyl-2-nitrophenol, methyl3-hydroxy-4-nitronemzoate
methyl 3-hydroxy-4-nitrobenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents