Please login to your online account to display your discounted pricing

Methyl 3-hydroxy-4-methoxybenzoate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 3-hydroxy-4-methoxybenzoate
Assay Percent Range 98%
Molecular Formula C9H10O4
Linear Formula CH3OC6H3(OH)COOCH3
Formula Weight 182.18
MDL Number MFCD01321262
Synonym Methyl isovanillate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC399260010

  SDS
thermo fisher scientific
AC399260010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC399260050

  SDS
thermo fisher scientific
AC399260050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 182.18
Quantity 1g
Synonym Methyl isovanillate
Chemical Name or Material Methyl 3-hydroxy-4-methoxybenzoate
Molecular Formula C9H10O4
CAS 6702-50-7
Assay Percent Range 98%
Linear Formula CH3OC6H3(OH)COOCH3
MDL Number MFCD01321262