missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-hydroxy-4-iodobenzoate, 97%

$48.58 - $575.22

Chemical Identifiers

CAS 157942-12-6
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD06203960
InChI Key LXCQVWRESZDFGW-UHFFFAOYSA-N
Synonym methyl-4-iodo-3-hydroxy benzoate, 3-hydroxy-4-iodobenzoic acid methyl ester, 3-hydroxy-4-iodo-benzoic acid methyl ester, methyl 3-hydroxy-4-iodobenzenecarboxylate, benzoic acid, 3-hydroxy-4-iodo-, methyl ester, rarechem al bf 0964, methyl 3-hydroxy4-iodobenzoate, methyl 4-iodo-3-hydroxybenzoate, methyl-3-hydroxy-4-iodobenzoate, methyl-4-iodo-3-hydroxybenzoate
PubChem CID 10378812
IUPAC Name methyl 3-hydroxy-4-iodobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)I)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64354MD View Documents Alfa Aesar™
H64354MD
250mg Each for $48.58
 
AAH6435403 View Documents Alfa Aesar™
H6435403
1g Each for $144.34
 
AAH6435406 View Documents Alfa Aesar™
H6435406
5g Each for $575.22
 
Description

Description

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
Specifications

Chemical Identifiers

157942-12-6
278.045
LXCQVWRESZDFGW-UHFFFAOYSA-N
10378812
COC(=O)C1=CC(=C(C=C1)I)O
C8H7IO3
MFCD06203960
methyl-4-iodo-3-hydroxy benzoate, 3-hydroxy-4-iodobenzoic acid methyl ester, 3-hydroxy-4-iodo-benzoic acid methyl ester, methyl 3-hydroxy-4-iodobenzenecarboxylate, benzoic acid, 3-hydroxy-4-iodo-, methyl ester, rarechem al bf 0964, methyl 3-hydroxy4-iodobenzoate, methyl 4-iodo-3-hydroxybenzoate, methyl-3-hydroxy-4-iodobenzoate, methyl-4-iodo-3-hydroxybenzoate
methyl 3-hydroxy-4-iodobenzoate

Specifications

Light Sensitive
157942-12-6
MFCD06203960
methyl-4-iodo-3-hydroxy benzoate, 3-hydroxy-4-iodobenzoic acid methyl ester, 3-hydroxy-4-iodo-benzoic acid methyl ester, methyl 3-hydroxy-4-iodobenzenecarboxylate, benzoic acid, 3-hydroxy-4-iodo-, methyl ester, rarechem al bf 0964, methyl 3-hydroxy4-iodobenzoate, methyl 4-iodo-3-hydroxybenzoate, methyl-3-hydroxy-4-iodobenzoate, methyl-4-iodo-3-hydroxybenzoate
COC(=O)C1=CC(=C(C=C1)I)O
278.045
278.05
97%
250mg
C8H7IO3
7289690
LXCQVWRESZDFGW-UHFFFAOYSA-N
methyl 3-hydroxy-4-iodobenzoate
10378812
Solid
Methyl 3-hydroxy-4-iodobenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning