missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-Hydroxy-2-pyridinecarboxylate, TCI America™

$120.70 - $424.73

Chemical Identifiers

CAS 62733-99-7
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00661311
InChI Key MHKKUZDJUGIOBC-UHFFFAOYSA-N
Synonym 3-Hydroxy-2-pyridinecarboxylic Acid Methyl Ester, Methyl 3-Hydroxypicolinate, 3-Hydroxypicolinic Acid Methyl Ester
PubChem CID 819341
IUPAC Name methyl 3-hydroxypyridine-2-carboxylate
SMILES COC(=O)C1=C(C=CC=N1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M29731G View Documents TCI America
M29731G
1g Each for $120.70
 
M29735G View Documents TCI America
M29735G
5g Each for $424.73
 
Specifications

Chemical Identifiers

62733-99-7
153.137
MHKKUZDJUGIOBC-UHFFFAOYSA-N
819341
COC(=O)C1=C(C=CC=N1)O
C7H7NO3
MFCD00661311
3-Hydroxy-2-pyridinecarboxylic Acid Methyl Ester, Methyl 3-Hydroxypicolinate, 3-Hydroxypicolinic Acid Methyl Ester
methyl 3-hydroxypyridine-2-carboxylate

Specifications

White-Yellow
1g
C7H7NO3
3-Hydroxy-2-pyridinecarboxylic Acid Methyl Ester, Methyl 3-Hydroxypicolinate, 3-Hydroxypicolinic Acid Methyl Ester
COC(=O)C1=C(C=CC=N1)O
153.137
153.14
≥98.0% (GC)
77°C
62733-99-7
MFCD00661311
MHKKUZDJUGIOBC-UHFFFAOYSA-N
methyl 3-hydroxypyridine-2-carboxylate
819341
Crystal-Powder at 20°C
Methyl 3-Hydroxy-2-pyridinecarboxylate
Safety and Handling

Safety and Handling

TSCA : No