missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-Fluorophenylacetate, TCI America™

$100.83 - $351.39

Chemical Identifiers

CAS 64123-77-9
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD03093288
InChI Key OXYLCGRXCQARQV-UHFFFAOYSA-N
Synonym methyl 2-3-fluorophenyl acetate, methyl 3-fluorophenylacetate, methyl3-fluorophenylacetate, methyl 3-fluorophenyl acetate, 3-fluorobenzeneacetic acid methyl ester, benzeneacetic acid, 3-fluoro-, methyl ester, 3-fluorophenylacetic acid methyl ester, 3-fluorophenyl acetic acid methyl ester, 3-fluoro-phenyl-acetic acid methyl ester, 2-3-fluorophenyl acetic acid methyl ester
PubChem CID 2734108
IUPAC Name methyl 2-(3-fluorophenyl)acetate
SMILES COC(=O)CC1=CC(=CC=C1)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M253625G View Documents TCI America
M253625G
25g Each for $351.39
 
M25365G View Documents TCI America
M25365G
5g Each for $100.83
 
Specifications

Chemical Identifiers

64123-77-9
168.167
OXYLCGRXCQARQV-UHFFFAOYSA-N
2734108
COC(=O)CC1=CC(=CC=C1)F
C9H9FO2
MFCD03093288
methyl 2-3-fluorophenyl acetate, methyl 3-fluorophenylacetate, methyl3-fluorophenylacetate, methyl 3-fluorophenyl acetate, 3-fluorobenzeneacetic acid methyl ester, benzeneacetic acid, 3-fluoro-, methyl ester, 3-fluorophenylacetic acid methyl ester, 3-fluorophenyl acetic acid methyl ester, 3-fluoro-phenyl-acetic acid methyl ester, 2-3-fluorophenyl acetic acid methyl ester
methyl 2-(3-fluorophenyl)acetate

Specifications

Colorless
64123-77-9
MFCD03093288
OXYLCGRXCQARQV-UHFFFAOYSA-N
methyl 2-(3-fluorophenyl)acetate
2734108
Clear Liquid at 20°C
Methyl 3-Fluorophenylacetate
25g
C9H9FO2
methyl 2-3-fluorophenyl acetate, methyl 3-fluorophenylacetate, methyl3-fluorophenylacetate, methyl 3-fluorophenyl acetate, 3-fluorobenzeneacetic acid methyl ester, benzeneacetic acid, 3-fluoro-, methyl ester, 3-fluorophenylacetic acid methyl ester, 3-fluorophenyl acetic acid methyl ester, 3-fluoro-phenyl-acetic acid methyl ester, 2-3-fluorophenyl acetic acid methyl ester
COC(=O)CC1=CC(=CC=C1)F
168.167
168.17
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No