Please login to your online account to display your discounted pricing

Methyl 3-fluorophenylacetate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  351090050

 View more versions of this product

Catalog No. AC351090050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 168.17
Quantity 5g
Color Clear colorless to slightly yellow
Chemical Name or Material Methyl 3-fluorophenylacetate
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4870 to 1.4890 (20°C, 589nm)
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9H9FO2
Linear Formula FC6H4CHCOH
MDL Number MFCD03093288
CAS 64123-77-9

Specifications

Chemical Name or Material Methyl 3-fluorophenylacetate
Name Note 98%
CAS 64123-77-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9FO2
Linear Formula FC6H4CHCOH
Formula Weight 168.17
MDL Number MFCD03093288