missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-fluoro-4-hydroxybenzoate, 98%

$30.70 - $394.78

Chemical Identifiers

CAS 403-01-0
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD00215838
InChI Key IYUSGKSCDUJSKS-UHFFFAOYSA-N
Synonym 3-fluoro-4-hydroxy benzoic acid methyl ester
PubChem CID 7019368
IUPAC Name methyl 3-fluoro-4-hydroxybenzoate
SMILES COC(=O)C1=CC(=C(C=C1)O)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61219MD View Documents Alfa Aesar™
H61219MD
250mg Each for $30.70
 
AAH6121903 View Documents Alfa Aesar™
H6121903
1g Each for $90.22
 
AAH6121906 View Documents Alfa Aesar™
H6121906
5g Each for $394.78
 
Specifications

Chemical Identifiers

403-01-0
170.139
IYUSGKSCDUJSKS-UHFFFAOYSA-N
7019368
COC(=O)C1=CC(=C(C=C1)O)F
C8H7FO3
MFCD00215838
3-fluoro-4-hydroxy benzoic acid methyl ester
methyl 3-fluoro-4-hydroxybenzoate

Specifications

250mg
C8H7FO3
3-fluoro-4-hydroxy benzoic acid methyl ester
COC(=O)C1=CC(=C(C=C1)O)F
170.139
170.14
98%
403-01-0
MFCD00215838
IYUSGKSCDUJSKS-UHFFFAOYSA-N
methyl 3-fluoro-4-hydroxybenzoate
7019368
Solid
Methyl 3-fluoro-4-hydroxybenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning