Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-dimethylamino-2-methylpropionate, 98%

$59.41 - $891.24

Chemical Identifiers

Chemical Name or Material Methyl 3-dimethylamino-2-methylpropionate
CAS 10205-34-2
Molecular Formula C7H15NO2
Formula Weight 145.2
MDL Number MFCD00044842
Beilstein 1755266
Synonym 3-Dimethylamino-2-methylpropionic acid methyl ester; Methyl beta-(dimethylamino)isobutyrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1903409

 
alfa aesar™
A1903409
10g Each for $59.41

AAA1903430

 
alfa aesar™
A1903430
250g Each for $891.24
Description & Specifications

Specifications

Formula Weight 145.2
Chemical Name or Material Methyl 3-dimethylamino-2-methylpropionate
Assay 98%
Molecular Formula C7H15NO2
MDL Number MFCD00044842
CAS 10205-34-2
Beilstein 1755266
Synonym 3-Dimethylamino-2-methylpropionic acid methyl ester; Methyl beta-(dimethylamino)isobutyrate
Health Hazard 1 Warning
Boiling Point 152° to 154°C
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.