missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-Cyclopropyl-3-oxopropionate, TCI America™

$44.31 - $132.92

Chemical Identifiers

CAS 32249-35-7
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00274090
InChI Key RIJWDPRXCXJDPK-UHFFFAOYSA-N
Synonym 3-Cyclopropyl-3-oxopropionic Acid Methyl Ester
PubChem CID 10130016
IUPAC Name methyl 3-cyclopropyl-3-oxopropanoate
SMILES COC(=O)CC(=O)C1CC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M227725G View Documents TCI America
M227725G
25g Each for $132.92
 
M22775G View Documents TCI America
M22775G
5g Each for $44.31
 
Specifications

Chemical Identifiers

32249-35-7
142.154
RIJWDPRXCXJDPK-UHFFFAOYSA-N
10130016
COC(=O)CC(=O)C1CC1
C7H10O3
MFCD00274090
3-Cyclopropyl-3-oxopropionic Acid Methyl Ester
methyl 3-cyclopropyl-3-oxopropanoate

Specifications

80°C
25g
C7H10O3
3-Cyclopropyl-3-oxopropionic Acid Methyl Ester
COC(=O)CC(=O)C1CC1
142.154
142.15
≥96.0% (GC)
Yellow
32249-35-7
MFCD00274090
RIJWDPRXCXJDPK-UHFFFAOYSA-N
methyl 3-cyclopropyl-3-oxopropanoate
10130016
Clear Liquid at 20°C
Methyl 3-Cyclopropyl-3-oxopropionate
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4627

TSCA : No