missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-cyanocinnamate

$34.64 - $48.49

Chemical Identifiers

CAS 52116-81-1
Molecular Formula C11H9NO2
Formula Weight 187.2
MDL Number MFCD08064268
Synonym 3-Cyanocinnamic acid methyl ester; Methyl 3-(3-cyanophenyl)acrylate
Chemical Name or Material Methyl 3-cyanocinnamate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5087203 SDS Alfa Aesar™
H5087203
1g Each for $48.49
AAH50872MD SDS Alfa Aesar™
H50872MD
250mg Each for $34.64
Specifications Description & Specifications

Chemical Identifiers

52116-81-1
187.2
3-Cyanocinnamic acid methyl ester; Methyl 3-(3-cyanophenyl)acrylate
C11H9NO2
MFCD08064268
Methyl 3-cyanocinnamate

Specifications

1g
C11H9NO2
MFCD08064268
Methyl 3-cyanocinnamate
52116-81-1
187.2
3-Cyanocinnamic acid methyl ester; Methyl 3-(3-cyanophenyl)acrylate

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger