Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-chlorothiophene-2-carboxylate, 97%

$73.08 - $73.08

Chemical Identifiers

Chemical Name or Material Methyl 3-chlorothiophene-2-carboxylate
CAS 88105-17-3
Molecular Formula C6H5ClO2S
Formula Weight 176.62
MDL Number MFCD00068135
Synonym 3-Chlorothiophene-2-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2022403

  SDS
alfa aesar™
B2022403
1g Each for $73.08
Description & Specifications

Specifications

Formula Weight 176.62
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 72° to 74°C (0.2mmHg)
CAS 88105-17-3
MDL Number MFCD00068135
Molecular Formula C6H5ClO2S
Assay 97%
Synonym 3-Chlorothiophene-2-carboxylic acid methyl ester
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Methyl 3-chlorothiophene-2-carboxylate
Melting Point 37° to 38°C
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.