missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 3-chlorophenylacetate, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 53088-68-9 |
---|---|
Molecular Formula | C9H9ClO2 |
Molecular Weight (g/mol) | 184.62 |
MDL Number | MFCD00800679 |
InChI Key | HTBBVKFFDDQVER-UHFFFAOYSA-N |
Synonym | methyl 2-3-chlorophenyl acetate, methyl 3-chlorophenylacetate, methyl 3-chlorophenyl acetate, phenylacetic acid, 3-chloro, methyl ester, benzeneacetic acid, 3-chloro-, methyl ester, methyl2-3-chlorophenyl acetate, methyl 2-3-chlorophenyl ethanoate, 3-chlorophenylacetic acid methyl ester, 3-chlorophenyl acetic acid methyl ester |
PubChem CID | 2733544 |
IUPAC Name | methyl 2-(3-chlorophenyl)acetate |
SMILES | COC(=O)CC1=CC=CC(Cl)=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC371670250
|
Thermo Scientific Chemicals
371670250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
53088-68-9 | |
184.62 | |
HTBBVKFFDDQVER-UHFFFAOYSA-N | |
2733544 | |
COC(=O)CC1=CC=CC(Cl)=C1 |
C9H9ClO2 | |
MFCD00800679 | |
methyl 2-3-chlorophenyl acetate, methyl 3-chlorophenylacetate, methyl 3-chlorophenyl acetate, phenylacetic acid, 3-chloro, methyl ester, benzeneacetic acid, 3-chloro-, methyl ester, methyl2-3-chlorophenyl acetate, methyl 2-3-chlorophenyl ethanoate, 3-chlorophenylacetic acid methyl ester, 3-chlorophenyl acetic acid methyl ester | |
methyl 2-(3-chlorophenyl)acetate |
Specifications
53088-68-9 | |
Authentic | |
Glass bottle | |
ClC6H4CH2C(O)OCH3 | |
MFCD00800679 | |
1.2 | |
HTBBVKFFDDQVER-UHFFFAOYSA-N | |
methyl 2-(3-chlorophenyl)acetate | |
2733544 | |
Liquid | |
Methyl 3-chlorophenylacetate |
1.2000g/mL | |
97.5% min. (GC) | |
C9H9ClO2 | |
1.5225 to 1.5245 | |
25g | |
methyl 2-3-chlorophenyl acetate, methyl 3-chlorophenylacetate, methyl 3-chlorophenyl acetate, phenylacetic acid, 3-chloro, methyl ester, benzeneacetic acid, 3-chloro-, methyl ester, methyl2-3-chlorophenyl acetate, methyl 2-3-chlorophenyl ethanoate, 3-chlorophenylacetic acid methyl ester, 3-chlorophenyl acetic acid methyl ester | |
COC(=O)CC1=CC=CC(Cl)=C1 | |
184.62 | |
184.51 | |
98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO â Research Use Only