Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-chlorobenzo[b]thiophene-2-carboxylate, 95%

$206.43 - $206.43

Chemical Identifiers

Chemical Name or Material Methyl 3-chlorobenzo[b]thiophene-2-carboxylate
CAS 21211-07-4
Molecular Formula C10H7ClO2S
Formula Weight 226.68
MDL Number MFCD00052503
Beilstein 1312686
Synonym 3-Chlorobenzo[b]thiophene-2-carboxylic acid methyl ester; Methyl 3-chlorothianaphthene-2-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1069606

  SDS
alfa aesar™
L1069606
5g Each for $206.43
Description & Specifications

Specifications

Formula Weight 226.68
CAS 21211-07-4
Chemical Name or Material Methyl 3-chlorobenzo[b]thiophene-2-carboxylate
Assay 95%
Molecular Formula C10H7ClO2S
Beilstein 1312686
Synonym 3-Chlorobenzo[b]thiophene-2-carboxylic acid methyl ester; Methyl 3-chlorothianaphthene-2-carboxylate
Melting Point 80° to 82°C
Quantity 5g
Odor Odorless
MDL Number MFCD00052503