missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-chlorobenzo[b]thiophene-2-carboxylate, 95%, Alfa Aesar™

Manufacturer:  Alfa Aesar L1069606

 View more versions of this product

Catalog No. AAL1069606



Specifications

Specifications

Methyl 3-chlorobenzo[b]thiophene-2-carboxylate
80°C to 82°C
21211-07-4
MFCD00052503
methyl 3-chlorobenzo b thiophene-2-carboxylate, benzo b thiophene-2-carboxylic acid, 3-chloro-, methyl ester, methyl 3-chlorobenzo b thiophene-2-carboxylate, maybridge1_000462, acmc-1co9m, methyl 3-chlorobenzothiophene-2-carboxylate, methyl 3-chloranyl-1-benzothiophene-2-carboxylate, methyl 3-chlorobenzo b thiophene-2-carboxylate, 3-chloro-1-benzothiophene-2-carboxylic acid methyl ester
COC(=O)C1=C(C2=CC=CC=C2S1)Cl
226.674
226.68
Odorless
5g
C10H7ClO2S
1312686
VAPMGETZRJYSLW-UHFFFAOYSA-N
methyl 3-chloro-1-benzothiophene-2-carboxylate
607073
95%

Chemical Identifiers

21211-07-4
226.674
VAPMGETZRJYSLW-UHFFFAOYSA-N
607073
COC(=O)C1=C(C2=CC=CC=C2S1)Cl
C10H7ClO2S
MFCD00052503
methyl 3-chlorobenzo b thiophene-2-carboxylate, benzo b thiophene-2-carboxylic acid, 3-chloro-, methyl ester, methyl 3-chlorobenzo b thiophene-2-carboxylate, maybridge1_000462, acmc-1co9m, methyl 3-chlorobenzothiophene-2-carboxylate, methyl 3-chloranyl-1-benzothiophene-2-carboxylate, methyl 3-chlorobenzo b thiophene-2-carboxylate, 3-chloro-1-benzothiophene-2-carboxylic acid methyl ester
methyl 3-chloro-1-benzothiophene-2-carboxylate
Documents