missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-Chloro-4-methylbenzoate, TCI America™

$79.45 - $239.86

Chemical Identifiers

CAS 56525-63-4
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00205141
InChI Key KTFQDZCNPGFKAH-UHFFFAOYSA-N
Synonym methyl3-chloro-4-methylbenzoate, methyl-3-chloro-4-methylbenzoate, methyl 3-chloro-p-toluate, 2-chloro-4-methoxycarbonyl toluene, 3-chloro-4-methylbenzoic acid methyl ester, benzoic acid, 3-chloro-4-methyl-, methyl ester, pubchem10935, acmc-209lsv, ksc495i9j, methyl-3-chloro-4-metylbenzoate
PubChem CID 2801405
IUPAC Name methyl 3-chloro-4-methylbenzoate
SMILES CC1=C(C=C(C=C1)C(=O)OC)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M21365G View Documents TCI America
M21365G
5g Each for $79.45
 
M213625G View Documents TCI America
M213625G
25g Each for $239.86
 
Specifications

Chemical Identifiers

56525-63-4
184.619
KTFQDZCNPGFKAH-UHFFFAOYSA-N
2801405
CC1=C(C=C(C=C1)C(=O)OC)Cl
C9H9ClO2
MFCD00205141
methyl3-chloro-4-methylbenzoate, methyl-3-chloro-4-methylbenzoate, methyl 3-chloro-p-toluate, 2-chloro-4-methoxycarbonyl toluene, 3-chloro-4-methylbenzoic acid methyl ester, benzoic acid, 3-chloro-4-methyl-, methyl ester, pubchem10935, acmc-209lsv, ksc495i9j, methyl-3-chloro-4-metylbenzoate
methyl 3-chloro-4-methylbenzoate

Specifications

107°C
5g
C9H9ClO2
methyl3-chloro-4-methylbenzoate, methyl-3-chloro-4-methylbenzoate, methyl 3-chloro-p-toluate, 2-chloro-4-methoxycarbonyl toluene, 3-chloro-4-methylbenzoic acid methyl ester, benzoic acid, 3-chloro-4-methyl-, methyl ester, pubchem10935, acmc-209lsv, ksc495i9j, methyl-3-chloro-4-metylbenzoate
CC1=C(C=C(C=C1)C(=O)OC)Cl
184.619
184.62
≥98.0% (GC)
White-Yellow
56525-63-4
MFCD00205141
KTFQDZCNPGFKAH-UHFFFAOYSA-N
methyl 3-chloro-4-methylbenzoate
2801405
Crystal-Lump at 20°C
Methyl 3-Chloro-4-methylbenzoate
Safety and Handling

Safety and Handling

TSCA : No