Please login to your online account to display your discounted pricing

Methyl 3-chloro-4-methylbenzoate, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 3-chloro-4-methylbenzoate
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H9ClO2
Formula Weight 184.62
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00703DA

 
fisher scientific company
TL00703DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 184.62
Quantity 1g
Assay 97%
Chemical Name or Material Methyl 3-chloro-4-methylbenzoate
Assay Percent Range 97%
Molecular Formula C9H9ClO2
CAS 56525-63-4