Please login to your online account to display your discounted pricing

Methyl 3-chloro-4-methylbenzoate, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  TL00703DA

 View more versions of this product

Catalog No. TL00703DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 184.62
Quantity 1g
Assay 97%
Chemical Name or Material Methyl 3-chloro-4-methylbenzoate
Molecular Formula C9H9ClO2
CAS 56525-63-4
Assay Percent Range 97%

Specifications

Chemical Name or Material Methyl 3-chloro-4-methylbenzoate
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H9ClO2
Formula Weight 184.62