Please login to your online account to display your discounted pricing

Methyl 3-chloro-4-methoxybenzoate 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 3-chloro-4-methoxybenzoate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9ClO3
Linear Formula CH3OC6H3(Cl)COOCH3
Formula Weight 200.62
MDL Number MFCD00017188
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC381880500

  SDS
thermo fisher scientific
AC381880500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 200.62
Quantity 50g
Color White to light yellow
Melting Point 93° to 97°C
Assay Percent Range 98%
Molecular Formula C9H9ClO3
Linear Formula CH3OC6H3(Cl)COOCH3
MDL Number MFCD00017188
Packaging Glass bottle
Chemical Name or Material Methyl 3-chloro-4-methoxybenzoate
CAS 37908-98-8
Infrared Spectrum Authentic