Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-chloro-4-methoxybenzoate, 98%

$96.51 - $1,317.62

Chemical Identifiers

Chemical Name or Material Methyl 3-chloro-4-methoxybenzoate
CAS 37908-98-8
Molecular Formula C9H9ClO3
Formula Weight 200.62
MDL Number MFCD00017188
Beilstein 2365816
Synonym 3-Chloro-4-methoxybenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1335609

 
alfa aesar™
A1335609
10g Each for $96.51

AAA1335630

 
alfa aesar™
A1335630
250g Each for $1,317.62
Description & Specifications

Specifications

Formula Weight 200.62
Chemical Name or Material Methyl 3-chloro-4-methoxybenzoate
Assay 98%
Molecular Formula C9H9ClO3
MDL Number MFCD00017188
CAS 37908-98-8
Beilstein 2365816
Synonym 3-Chloro-4-methoxybenzoic acid methyl ester
Melting Point 93° to 97°C
Quantity 10g